Learn More
Fisherbrand™ Dust'it™ Air Duster
Powerful bulb duster forcibly blows away dust, lint and dirt
Marca:  Fisherbrand™ 1464959
Código de producto. 18874721
  • 159.00€ / paquete de 3
Fecha estimada de envío: 20-06-2025
para ver el stock
Explore más ofertas especiales
Descripción
Powerful bulb duster forcibly blows away dust, lint and dirt
Ideal for optical lenses, slides, circuit boards and cuvets.
  • Provides high-pressure, pinpoint jets of air exactly where desired; vary squeeze to control delivery
  • One-way inlet valve at top stops air from being drawn through tip
  • Nonconductive plastic sleeve-tip prevents scratches
Especificaciones
Especificaciones
Air Duster
None
Aerosol
Unscented
Eliminates dust, lint and dirt; cleans optical lenses, slides and other delicate, difficult-to-clean and inaccessible equipment
56.7 g
None
No
None
No estéril
2 oz.
Certificados
Ofertas especiales
Ofertas especiales
Título del producto
Fisherbrand™ Dust'it™ Air Duster > Dust'it Air Duster