Learn More
Fisherbrand™ Autoclavable HD Biohazard Waste Bags
Strong and leak-resistant
Marca: Fisherbrand™ 22044562
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Strong and leak-resistant
- Printed with biohazard symbol and text in English and Spanish
- Supplied with twist tie closure
Especificaciones
Twist Tie | |
19 x 14 in. (48 x 36 cm) | |
36 cm | |
Biohazard symbol and text in English and Spanish | |
19 in. | |
Polyethylene |
Red with Black Print | |
48 cm | |
Autoclavable | |
14 in. | |
1.25 mil |