missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FITC Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marca: Invitrogen PA126793
Detalles adicionales : Peso : 1.24740kg
Descripción
PA1-26793 detects Fluorescein.
FITC (fluorescein isothiocyanate) is a fluorochrome dye that absorbs μgtraviolet or blue light causing molecules to become excited and emit a visible yellow-green light. This emission ceases upon removal of the light causing the excitation. Fluorochrome labeling provides rapid, accurate localization of antigen-antibody interaction when one of the reactants is part of a cell, tissue or other biological structure. FITC is a commonly used marker for antibodies in immunofluorescent techniques since the conjugation of FITC to proteins is relatively easy and does not, in general, destroy the biological activity of the labeled protein. FITC is widely used as a hapten to label different proteins. Anti-FITC antibodies can be used for isolating FITC labeled cells by magnetic microbeads based sorting, as well as removal of background signal.Especificaciones
FITC | |
Polyclonal | |
Unconjugated | |
Fluorescein | |
Fluorescein, conjugated to goat IgG. | |
100 μg | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
Chemical |
ELISA, Immunodiffusion, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
1.2 mg/mL | |
0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
Chemical | |
Antibody | |
IgG |