missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FJX1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA542965
Detalles adicionales : Peso : 0.00010kg
Descripción
Peptide sequence: MVALERGGCG RSSNRLARFA DGTRACVRYG INPEQIQGEA LSYYLARLLG Sequence homology: Dog: 79%; Guinea Pig: 100%; Human: 100%; Mouse: 100%; Rat: 100%
FJX1 Polyclonal antibody specifically detects FJX1 in Human samples. It is validated for Western Blot.Especificaciones
FJX1 | |
Polyclonal | |
Unconjugated | |
FJX1 | |
FJX1; FLJ22416; FLJ25593; four jointed box 1; four jointed box 1 (Drosophila); four-jointed box kinase 1; four-jointed box protein 1; Four-jointed protein homolog; four-jointed protein x1; LOW QUALITY PROTEIN: four-jointed box protein 1; putative secreted ligand homologous to fjx1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
24147 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q86VR8 | |
FJX1 | |
Synthetic peptide directed towards the N-terminal of human FJX1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |