Learn More
FKBP14 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA555430
Detalles adicionales : Peso : 0.02000kg
Descripción
Immunogen sequence: ALIPEPEVKI EVLQKPFICH RKTKGGDLML VHYEGYLEKD GSLFHSTHKH NNGQPIWFTL GILEALKGWD QGLKGMCVG Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.
PPIases accelerate the folding of proteins during protein synthesis.Especificaciones
FKBP14 | |
Polyclonal | |
Unconjugated | |
Fkbp14 | |
22 kDa FK506-binding protein; 22 kDa FKBP; BC029109; EDSKMH; FK506 binding protein 14; FK506 binding protein 14, 22 kDa; FK506-binding protein 14; Fkbp14; FKBP-14; FKBP-2; FKBP22; FKBP-22; IPBP12; peptidyl-prolyl cis-trans isomerase FKBP14; PPIase; PPIase FKBP14; rotamase; UNQ322/PRO381 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55033 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NWM8 | |
Fkbp14 | |
Recombinant protein corresponding to Human FKBP14 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only