Learn More
FKBP4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA522277
Detalles adicionales : Peso : 0.01000kg
Descripción
Recommended positive controls: 293T, A431, HeLa, HepG2, A375, GL261, C8D30, Rat2. Predicted reactivity: Mouse (95%), Rat (94%), Pig (97%), Rabbit (93%), Chicken (85%), Rhesus Monkey (98%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The function of this protein remains unknown.Especificaciones
FKBP4 | |
Polyclonal | |
Unconjugated | |
FKBP4 | |
51 kDa FK506-binding protein; 52 kDa FK506-binding protein; 52 kDa FKBP; 59 kDa immunophilin; 59kDa; AL022792; AW208983; FK506 binding protein 4; FK506 binding protein 4, 59kDa; FK506 binding protein 52; FK506-binding protein 4; FK506-binding protein 4 (59kD); FKBP prolyl isomerase 4; Fkbp4; FKBP-4; FKBP51; FKBP52; FKBP-52; FKBP52 protein; FKBP59; Fkpb52; HBI; HSP binding immunophilin; Hsp56; hsp90 binding protein; HSP-binding immunophilin; immunophilin FKBP52; p52; P59; p59 protein; peptidylprolyl cis-trans isomerase; peptidyl-prolyl cis-trans isomerase FKBP4; Peptidyl-prolyl cis-trans isomerase FKBP4, N-terminally processed; PPIase; PPIase FKBP4; rotamase; T-cell FK506-binding protein, 59kD | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
14228, 2288, 260321 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
0.99 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
P30416, Q02790, Q9QVC8 | |
FKBP4 | |
Recombinant fragment corresponding to a region within amino acids 1 and 207 of FKBP52 (Uniprot ID#Q02790) | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |