Learn More
FKRP Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA563857
Detalles adicionales : Peso : 0.02000kg
Descripción
Immunogen sequence: ETARYVVGVL EAAGVRYWLE GGSLLGAARH GDIIPWDYDV DLGIYLEDVG NCEQLRGAEA GSVVDERGFV WEKAVEGDF Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
This gene encodes a protein which is targeted to the medial Golgi apparatus and is necessary for posttranslational modification of dystroglycan. Mutations in this gene have been associated with congenital muscular dystrophy, mental retardation, and cerebellar cysts. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined. [provided by RefSeq, Oct 2008].Especificaciones
FKRP | |
Polyclonal | |
Unconjugated | |
FKRP | |
A830029B19Rik; AI842067; AI847300; FKRP; fukutin related protein; fukutin-related protein; LGMD1I; LGMD2I; MDC1C; MDDGA5; MDDGB5; MDDGC5; Ribitol-5-phosphate transferase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79147 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H9S5 | |
FKRP | |
Recombinant protein corresponding to Human FKRP | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only