Descripción
- Most economical solution without sacrificing quality for standard separations
- Large selection of cartridge sizes from 4g-5,000g
- Non-RFID suitable on any system from BUCHI, ISCO, Biotage, Interchim or Yamazen
- Silica is ideal for basic applications and polar compounds using organic solvents
- Amino functionalized silica provides better peak shapes with polar and basic compounds and is ideal for carbohydrates
- C18 functionalized silica is ideal for peptides and non-polar compounds in an aqueous mobile phase
- Diol functionalized silica provides an alternate selectivity for a variety of polarity types and is ideal for lipids
- Neutral Alumina is ideal for aldehydes, ketones, and esters
Especificaciones
Especificaciones
| Material | Polipropileno |
| Tipo | Sílice |
| Incluye | 12 unidades |
| Resolución | 40-63 μm |
| Peso del lecho (métrico) | 25 g |
| Tipo de columna | Sílice |
| Para utilizar con (aplicación) | Aplicaciones básicas, compuestos polares |
| Para utilizar con (equipo) | Any Flash System |