Learn More
Grant Instruments™ Flask Tray for Grant Shaking Water Baths
Include threaded holes to accept flask clamps or holder for deep well plates (=2ml). Grant Instruments™ Flask Tray for Grant Shaking Water Baths are compatible with SR plate holders and racks or can be used alone to accommodate bags and miscellaneous vessels.
284.00€ - 326.48€
Especificaciones
Capacidad (métrico) | De 25 ml a 1000 ml |
---|---|
Tipo | Bandeja para matraces |
Código de producto | Marca | Para utilizar con (equipo) | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Para utilizar con (equipo) | Precio | Cantidad y disponibilidad | |||||
15345822
|
Grant Instruments™
TF12 |
LSB12 |
284.00€
cada uno |
En stock
Inicie sesión para ver el stock |
|||||
15375822
|
Grant Instruments™
TF18 |
LSB18 |
326.48€
cada uno |
En stock
Inicie sesión para ver el stock |
|||||
Descripción
Include threaded holes to accept flask clamps or holder for deep well plates (=2ml). Grant Instruments™ Flask Tray for Grant Shaking Water Baths are compatible with SR plate holders and racks or can be used alone to accommodate bags and miscellaneous vessels.
- Accommodates flasks from 25 to 1000mL
- For use with the followng Shaking Water Bath models:
- LSB12
- LSB18
- OLS26
- Compatible with SH plate holders or SR racks
- TF26 Test tube tray also compatible with SR racks
- Can be used alone to accommodate bags and miscellaneous vessels.
Especificaciones
De 25 ml a 1000 ml | |
Bandeja para matraces |