Learn More
Proteintech FlexAble CoraLite Plus 555 Antibody Labeling Kit for Human IgG
FlexAble CoraLite Plus 555 Antibody Labeling Kit for Human IgG is a novel antibody labeling kit that uses an affinity linker to conjugate CoraLite Plus 555 in any buffer condition to Human IgG primary antibodies from any supplier.
Marca:  Proteintech KFA105-10
 Ver más versiones de este producto
Código de producto. 17981427
  • 90.00€ / paquete de 10
Explore más ofertas especiales
Descripción
Descripción
FlexAble CoraLite™ Plus 555 Antibody Labeling Kit for Human IgG is a novel antibody labeling kit that uses an affinity linker to conjugate CoraLite™ Plus 555 in any buffer condition to Human IgG primary antibodies from any supplier. One labeling reaction requires only 0.5 μg of antibody - regardless of the antibody concentration. Label your Human IgG antibody in<10 min without any additional equipment.
Especificaciones
Especificaciones
10 Tests