Learn More
Proteintech FlexAble CoraLite Plus 750 Antibody Labeling Kit for Human IgG
FlexAble CoraLite Plus 750 Antibody Labeling Kit for Human IgG is a novel antibody labeling kit that uses an affinity linker to conjugate CoraLite Plus 750 in any buffer condition to Human IgG primary antibodies from any supplier.
90.00€ - 889.00€
Especificaciones
Formato Liquid
Ver más especificaciones
Productos 3
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
18078065
Ver documentos
Proteintech
KFA107-50
50 Tests
419.00€
50 reacciones
18009855
Ver documentos
Proteintech
KFA107-200
100 Tests
889.00€
200 reacciones
18067265
Ver documentos
Proteintech
KFA107-10
10 Tests
90.00€
10 reacciones
Descripción
Descripción
FlexAble CoraLite™ Plus 750 Antibody Labeling Kit for Human IgG is a novel antibody labeling kit that uses an affinity linker to conjugate CoraLite™ Plus 750 in any buffer condition to Human IgG primary antibodies from any supplier. One labeling reaction requires only 0.5 μg of antibody - regardless of the antibody concentration. Label your Human IgG antibody in<10 min without any additional equipment.