missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FLRT3 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marca: Invitrogen PA547400
Detalles adicionales : Peso : 0.00010kg
Descripción
Approximately 10% cross-reactivity with recombinant human (rh) FLRT1 is observed and 5% cross-reactivity with rhFLRT2 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
FLRT3 Polyclonal antibody specifically detects FLRT3 in Human samples. It is validated for Western Blot.Especificaciones
FLRT3 | |
Polyclonal | |
Unconjugated | |
FLRT3 | |
5530600M07Rik; C430047I10Rik; Fibronectin leucine rich transmembrane protein 3; fibronectin-like domain-containing leucine-rich transmembrane protein 3; FLRT3; HH21; KIAA1469; Leucine-rich repeat transmembrane protein FLRT3; mKIAA1469; RGD1566005; UNQ856/PRO1865 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
23767 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q9NZU0 | |
FLRT3 | |
Mouse myeloma cell line NS0-derived recombinant human FLRT3 Lys29-Pro528 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |