Learn More
FLT3LG Monoclonal Antibody (40406), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA523826
Detalles adicionales : Peso : 0.00010kg
Descripción
In direct ELISAs, this antibody does not cross-react with recombinant mouse Flt-3 Ligand. Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
FLT3LG Monoclonal antibody specifically detects FLT3LG in Human samples. It is validated for Neutralization, Western Blot.Especificaciones
FLT3LG | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P49771 | |
FLT3LG | |
Sf 21-derived recombinant human Flt-3 Ligand Thr27-Pro185 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Neutralization, Western Blot | |
40406 | |
Unconjugated | |
FLT3LG | |
FL; Flt 3 ligand; Flt3; Flt3 L; Flt3 ligand; Flt3l; Flt3lg; fms related receptor tyrosine kinase 3 ligand; fms related tyrosine kinase 3 ligand; FMS-like tyrosine kinase 3 ligand; fms-related tyrosine kinase 3; fms-related tyrosine kinase 3 ligand; H-FLT-3-L; Ly72L; M-FLT-3-L; NEWGENE_2322792; SL cytokine | |
Mouse | |
Protein A/G | |
RUO | |
2323 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |