Learn More
FMC1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA561063
Detalles adicionales : Peso : 0.02000kg
Descripción
Immunogen sequence: MAALGSPSHT FRGLLRELRY LSAATGRPYR DTAAYRYLVK AFRAHRVTSE KLCRAQHE Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%.
FMC1 Polyclonal antibody specifically detects FMC1 in Human samples. It is validated for Immunohistochemistry.Especificaciones
FMC1 | |
Polyclonal | |
Unconjugated | |
FMC1 | |
ATP synthase assembly factor FMC1, mitochondrial; C7orf55; C7orf55-LUC7L2; C7orf55-LUC7L2 protein; C7orf55-LUC7L2 readthrough; FMC1; Formation of mitochondrial complex V assembly factor 1 homolog; HSPC268; Protein FMC1 homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100996928, 154791 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96HJ9 | |
C7orf55-LUC7L2, FMC1 | |
Recombinant protein corresponding to Human FMC1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only