Learn More
FMNL3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA551720
Detalles adicionales : Peso : 0.02000kg
Descripción
Immunogen sequence: LLDVKELGRG MELIRRECSI HDNSVLRNFL STNEGKLDKL QRDAKTAEEA YNAVVRYFGE SPKTTPPSVF FPVFVRFIRS YKEAEQENEA RKKQEEVMRE KQLAQEAKKL DAKTPSQRNK WQQQE Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
FMNL3, also known as Formin-like protein 3, is a 1028 amino acid containing protein involved in immunity or defense. This developmental protein is implicated in polarity control, invasion, migration, or metastasis through regulation of the Rho-related signaling pathway. It belongs to the formin homology family and contains a FH2 (formin homology 2) domain and a GBD/FH3 (Rho GTPase-binding/formin homology 3) domain and has high sequence identity to the mouse Wbp3 protein. Two different isoforms have been described. Itis suspectedbe involved in gastric cancer.Especificaciones
FMNL3 | |
Polyclonal | |
Unconjugated | |
FMNL3 | |
2700073B04Rik; FBP11; FHOD3; FMNL3; Formin homology 2 domain-containing protein 3; formin like 3; formin-like 3; formin-like 3 protein; formin-like protein 3; Frl2; KIAA2014; mKIAA2014; WBP3; WBP-3; WW domain binding protein 3; WW domain-binding protein 3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
91010 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IVF7 | |
FMNL3 | |
Recombinant protein corresponding to Human FMNL3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only