Learn More
FN3K Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA563951
Detalles adicionales : Peso : 0.02000kg
Descripción
Immunogen sequence: EVASLEALRS TGLVRVPRPM KVIDLPGGGA AFVMEHLKMK SLSSQASKLG EQM Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 60%.
FN3K catalyzes phosphorylation of fructosamines formed by glycation, the nonenzymatic reaction of glucose with primary amines followed by Amadori rearrangement. Phosphorylation of fructosamines may initiate metabolism of the modified amine and result in deglycation of glycated proteins.Especificaciones
FN3K | |
Polyclonal | |
Unconjugated | |
Fn3k | |
2310074G21Rik; FN3K; Fnsk; fructosamine 3 kinase; fructosamine 3-kinase; fructosamine-3-kinase; Protein-psicosamine 3-kinase FN3K; Protein-ribulosamine 3-kinase FN3K | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
64122 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H479 | |
Fn3k | |
Recombinant protein corresponding to Human FN3K | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only