Learn More
FNBP1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA528626
Detalles adicionales : Peso : 0.01000kg
Descripción
Recommended positive controls: H1299, NIH-3T3, JC, BCL-1, PC-12. Predicted reactivity: Mouse (100%), Rat (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is a member of the formin-binding-protein family. The protein contains an N-terminal Fer/Cdc42-interacting protein 4 (CIP4) homology (FCH) domain followed by a coiled-coil domain, a proline-rich motif, a second coiled-coil domain, a Rho family protein-binding domain (RBD), and a C-terminal SH3 domain. This protein binds sorting nexin 2 (SNX2), tankyrase (TNKS), and dynamin; an interaction between this protein and formin has not been demonstrated yet in human.Especificaciones
FNBP1 | |
Polyclonal | |
Unconjugated | |
FNBP1 | |
1110057E06Rik; 2210010H06Rik; FBP1; FBP17; Fnbp1; formin binding protein 1; formin binding protein 17; formin-binding protein 1; Formin-binding protein 17; hFBP17; Kiaa0554; MGC126804; Rapostlin | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
14269, 192348, 23048 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.92 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q80TY0, Q8R511, Q96RU3 | |
FNBP1 | |
Synthetic peptide corresponding to a region within amino acids 82 and 175 of Human FNBP1 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |