Learn More
FNBP4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA559035
Detalles adicionales : Peso : 0.02000kg
Descripción
Immunogen sequence: NGETAIGAEN SEKIDENSDK EMEVEESPEK IKVQTTPKVE EEQDLKFQIG ELANTLTSKF EFLGINRQSI SNFHVLLLQT ETRIAD Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 92%.
FNBP4 encodes a protein containing two tryptophan-rich WW domains that binds the proline-rich formin homology 1 domains of formin family proteins, suggesting a role in the regulation of cytoskeletal dynamics during cell division and migration. It also binds intersectin family proteins suggesting a role in the maintenance of membrane curvature at sites of nascent vesicle formation. Naturally occurring mutations in this gene are associated with Waardenburg anophthalmia syndrome.Especificaciones
FNBP4 | |
Polyclonal | |
Unconjugated | |
FNBP4 | |
FBP30; Fnbp30; Fnbp4; formin binding protein 30; formin binding protein 4; formin-binding protein 30; Formin-binding protein 4; Kiaa1014; mKIAA1014 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23360 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N3X1 | |
FNBP4 | |
Recombinant protein corresponding to Human FNBP4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only