missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXA1 Monoclonal Antibody (3A8), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA1091
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
MA1-091 has been successfully used in Western blot and immunohistochemistry (paraffin) applications with human, mouse and canine samples.
This gene encodes a member of the forkhead class of DNA-binding proteins. These hepatocyte nuclear factors are transcriptional activators for liver-specific transcripts such as albumin and transthyretin, and they also interact with chromatin. Similar family members in mice have roles in the regulation of metabolism and in the differentiation of the pancreas and liver.Especificaciones
FOXA1 | |
Monoclonal | |
1 mg/mL | |
PBS with 1mg/mL BSA, 30% glycerol and 0.05% sodium azide | |
P35582, P55317 | |
FOXA1 | |
Recombinant protein expressed in E.coli corresponding to amino acids 1-473 of human FOXA1. | |
100 μg | |
Primary | |
Canine, Human, Mouse | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
3A8 | |
Unconjugated | |
FOXA1 | |
Ci-fkh; fkd7; fkh; fork head domain; forkhead box A1; forkhead box protein A1; forkhead-7; Foxa1; FoxA4a; hepatocyte nuclear factor 3 alpha (winged helix transcription factor); hepatocyte nuclear factor 3, alpha; hepatocyte nuclear factor 3-alpha; HNF3A; HNF-3A; HNF3alpha; HNF-3-alpha; HNF3-alpha; MGC33105; MGC52590; MGC75967; pintallavis; TCF3A; Tcf-3a; tFoxA1; Transcription factor 3A; xfkh1; xfkh2 | |
Mouse | |
Protein A | |
RUO | |
15375, 3169, 490657 | |
-20°C | |
Liquid |