Learn More
OHAUS™ Frontier™ 5000 High Speed Centrifuge
Dependable high speed centrifuges, ideal for high-speed lab applications.
8355.00€ - 10435.00€
Especificaciones
Certificaciones/conformidad | CE, FCC |
---|---|
Refrigerado | Sí |
Pantalla | 2-line, Backlit LCD |
Sistema de accionamiento | Direct, Brushless Induction Drive |
Material | Corrosion-resistant Stainless Steel Chamber with Polycarbonate/ABS Plastic Lid |
Código de producto | Marca | Vel. máx | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Vel. máx | Precio | Cantidad y disponibilidad | |||||
30130229
|
OHAUS™
83041047 |
30,000 rpm |
10435.00€
cada uno |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
30130244
|
OHAUS™
83041045 |
20,000 rpm |
8355.00€
cada uno |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
Descripción
Powerful and Versatile Universal Centrifuges for Virtually Every Lab Application
Frontier™ 5000 Multi-Pro Centrifuges are designed for universal use in multiple applications in research, industrial, and clinical laboratories and can be combined with our range of rotors and accessories for customized use. Available in 9 models with capacities from 0,2 ml to 4x750 ml, they feature high-speed performance at maximum volume, automatic rotor identification, and safety features.
Unique Features:
- Our multi-purpose centrifuges offer a high-speed centrifugation platform which can be customized to fit workflow needs using a wide variety of rotors and accessories.
- The intuitive design of the centrifuges & accessories enable easy access to parameter settings, and quick rotation between applications. Features include automatic rotor recognition and backlit LCD.
- The performance of these centrifuges is propelled by German engineering, and high-quality components for reliable use. Constructed of chemical-resistant stainless steel to ensure durability in lab use.
- Frontier™ 5000 Series is equipped with protective features, including automatic over-speed protection, imbalance detection and self-diagnostic system to ensure reliable use and operator safety.
FC5720R (refrigerated) Centrifuge
- Compatible with up to 17 optional rotors and 90 accessories
- Capacities from microliter to 4x200 ml
- Improved sample processing through-put with additional rotor for blood collection tubes and standard conical tubes
- High-speed centrifugation: up to 20000 rpm for microtubes and up to 16000 rpm for 50 ml
- Temperature variable range of -20°C to 40°C
FC5830R (refrigerated) Centrifuge
p>
- Compatible with up to 11 optional rotors and 56 accessories
- Capacities from microliter to 6x250 ml
- Improved sample processing through-put with additional rotor for blood collection tubes and standard conical tubes
- High-speed centrifugation: up to 30000 rpm for microtubes and up to 21000 rpm for 50 ml
- Temperature variable range of -20°C to 40°C
Especificaciones
CE, FCC | |
2-line, Backlit LCD | |
Corrosion-resistant Stainless Steel Chamber with Polycarbonate/ABS Plastic Lid | |
≤60 ±2 dB(A) | |
No | |
-20°C to 40°C | |
2 Years |
Sí | |
Direct, Brushless Induction Drive | |
99 Programs | |
Yes | |
10 sec. to 99 hr. 59 min. 59 sec. | |
Centrífuga |