Learn More
Eurofins DiscoverX Fyn Protein, Active
Active, N-Terminal His6-tagged, recombinant, full-length, human Fyn, for use in Kinase Assays.
Marca: Eurofins DiscoverX 14-441M
Descripción
p59fyn is a member of the src-related gene family of non-receptor tyrosine kinases. Recent studies indicate that fyn kinase plays a critical role in mediating signal transduction through the T-cell receptor thereby initiating a signal transduction cascade leading to lymphokine secretion and cell proliferation. One of the earliest biochemical responses following T-cell receptor activation is an increase in tyrosine kinase activity and concomitant phosphorylation of several intracellular substrates. P59fyn kinase is an important mediator of this phosphorylation since it is physically associated with the T-cell receptor. Additional evidence supports the view that fyn participates in signal transduction pathways leading from multiple T-cell membrane proteins and hence may serve to integrate stimuli received from several independent receptor structures. In the case of the B-cell receptor, fyn is one of several kinases associated with the receptor and is activated following receptor cross-linking.
EMBL M14333
Especificaciones
mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 150 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 5% glycerol, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. | |
250 μg | |
Recombinant | |
Recombinant enzyme expressed in Sf21 insect cells | |
≥56% |
61.8 kDa | |
Human | |
N-terminal 6His | |
Full-length | |
Fyn, active |
El Programa Encompass de Fisher Scientific ofrece artículos que no forman parte de nuestra cartera de distribución. Estos productos no suelen tener imágenes ni descripciones detalladas. Sin embargo, nos comprometemos a mejorar su experiencia de compra. Utilice el siguiente formulario para hacernos llegar sus comentarios sobre el contenido de este producto.