missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GALR3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA533578
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Percent identity with other species by BLAST analysis: Human, Gorilla (100%) Monkey (88%) Marmoset, Rat, Elephant, Pig, Bloom alga (82%).
The neuropeptide galanin modulates a variety of physiologic processes including cognition/memory, sensory/pain processing, hormone secretion, and feeding behavior. The human galanin receptors are G protein-coupled receptors that functionally couple to their intracellular effector through distinct signaling pathways. GALR3 is found in many tissues and may be expressed as 1.4-, 2.4-, and 5-kb transcripts. GALR3 has been reported in brain, testis, adrenal gland, and pancreas. An EST has been isolated from an ovarian thecoma fibroma library.Especificaciones
GALR3 | |
Polyclonal | |
Unconjugated | |
GALR3 | |
Gal receptor-3; Gal3 receptor; GAL3-R; Galanin r3 receptor; galanin receptor 3; galanin receptor type 3; galanin receptor, family member 3; Galnr3; Galr3; Gal-r3; GALR-3; OTTHUMP00000028820 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
29235, 8484 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.3 mg/mL | |
PBS with 0.1% sodium azide | |
O60755, O88626 | |
GALR3 | |
Synthetic 17 amino acid peptide from 1st cytoplasmic domain of human GALR3. | |
50 μg | |
Primary | |
Human, Rat | |
Antibody | |
IgG |