missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Galunisertib

Click to view available options
Cantidad:
5 mg
25 mg
Tamaño de la unidad:
25 miligramos
5 miligramos
Descripción
Galunisertib is an ALK4 and ALK5 (TGFβRI) inhibitor (IC50 values are 77.7 and 172 nM, respectively). Also inhibits a range of other kinases with submicromolar IC50s including MINK, TGFβRII, ALK6 and ACVR2B. Inhibits TGFβ-induced SMAD phosphorylation in cancer cell lines in vitro and in vivo. Also reverses TGFβ-mediated suppression of T cell proliferation. Prevents TGFβ-mediated EMT and cell migration in vitro and exhibits antitumor efficacy in vivo. Used as component of growth media for urothelial organoids. Orally bioavailable.
Specifica
Specifica
| Nombre del producto químico o material | 4-[5,6-Dihydro-2-(6-methyl-2-pyridinyl)-4 H-pyrrolo[1,2-b]pyrazol-3-yl]-6-quinolinecarboxamide |
| CAS | 700874-72-2 |
| Cantidad | 25 mg |
| Objetivo | TGF-beta Receptor Inhibitors |
| Fórmula molecular | C22H19N5O |
| Formula Weight (peso de la fórmula) | 369.43 |
| Tipo de producto | Inhibitor |
| Pureza | 98% |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto