missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GDF10 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA511927
365.35 EUR hasta el 2025-03-29
Use el código promocional "24111" para aplicar la oferta.
Alertas:
Para beneficiarse del descuento, los clientes deben comprar tres unidades del mismo producto al precio de catálogo en un solo pedido para recibir el 33.33% de descuento No hay límite para los múltiplos de 3 que los clientes pueden comprar PCODE:24111
Descripción
The protein encoded by this gene is a member of the bone morphogenetic protein (BMP) family and the TGF-beta superfamily. This group of proteins is characterized by a polybasic proteolytic processing site which is cleaved to produce a mature protein containing seven conserved cysteine residues. The members of this family are regulators of cell growth and differentiation in both embryonic and adult tissues. Studies in mice suggest that the protein encoded by this gene plays a role in skeletal morphogenesis.
Especificaciones
GDF10 | |
Polyclonal | |
Unconjugated | |
GDF10 | |
BIP; Bmp3b; BMP-3B; Bone morphogenetic protein 3B; bone morphogenetic protein 3B; growth/differentiation factor 10; bone-inducing protein; Gdf10; GDF-10; growth differentiation factor 10; growth/differentiation factor 10; prepro bone inducing protein | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
2662 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
2 mg/mL | |
PBS with 0.09% sodium azide | |
P55107 | |
GDF10 | |
KLH conjugated synthetic peptide between 33-62 amino acids from the N-terminal region of human GDF10 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
GDF10 Polyclonal Antibody, Invitrogen™ > 400 μL; Unconjugated
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido