missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gelsolin Monoclonal Antibody (20), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen ABS0172002
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
ABS 017-20-02 detects Gelsolin from human samples. ABS 017-20-02 has been successfully used in ELISA and western blot procedures. ABS 017-20-02 does not work as capture antibody in sandwich ELISA. NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL
The protein encoded by this gene binds to the "e;plus"e; ends of actin monomers and filaments to prevent monomer exchange. The encoded calcium-regulated protein functions in both assembly and disassembly of actin filaments. Defects in this gene are a cause of familial amyloidosis Finnish type (FAF). Multiple transcript variants encoding several different isoforms have been found for this gene.Especificaciones
Gelsolin | |
Monoclonal | |
1.03 mg/mL | |
PBS with 15mM sodium azide; pH 7.4 | |
P06396 | |
GSN | |
Gelsolin purified from human plasma. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
20 | |
Unconjugated | |
GSN | |
Actin-depolymerizing factor; ADF; ADFBrevin; AGEL; Brevin; DKFZp313L0718; gelsolin; gelsolin (amyloidosis, Finnish type); Gsb; Gsn; hypothetical protein LOC535077; plasma gelsolin precursor; RP11-477J21.1 | |
Mouse | |
Protein A/G | |
RUO | |
2934 | |
4° C, store in dark | |
Liquid |