Learn More
Geminin Antibody (CPTC-GMNN-1), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NBP2-79878PCP
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Geminin Monoclonal antibody specifically detects Geminin in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Protein Array
Especificaciones
| Geminin | |
| Monoclonal | |
| PerCP | |
| Gem, geminin, geminin, DNA replication inhibitor, RP3-369A17.3 | |
| Recombinant human full-length protein (Uniprot: O75496) | |
| 0.1 mL | |
| Breast Cancer, Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, DNA Repair, DNA replication Transcription Translation and Splicing, Nuclear Receptors Coactivators and Corepressors, Stem Cell Markers | |
| 51053 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Protein Array | |
| CPTC-GMNN-1 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |