Learn More
Alla France™ Hidrómetro de densidad de uso general
Código de producto. 11788083
missing translation for 'orderingAttributeHoverText'
Intervalo de gravedad específica:
De 0,140 a 0,150 g/ml
De 0,150 a 0,160 g/ml
De 0,160 a 0,170 g/ml
De 0,170 a 0,180 g/ml
De 0,180 a 0,190 g/ml
De 0,600 a 0,700 g/ml
De 0,700 a 0,800 g/ml
De 0,800 a 0,900 g/ml
De 0,850 a 0,900 g/ml
De 0,900 a 0,950 g/ml
De 0,900 a 1,000 g/ml
De 1,050 a 1,100 g/ml
De 1,200 a 1,250 g/ml
De 1,250 a 1,300 g/ml
De 1,300 a 1,350 g/ml
De 1,300 a 1,400 g/ml
De 1,350 a 1,400 g/ml
De 1,400 a 1,450 g/ml
De 1,400 a 1,500 g/ml
De 1,500 a 1,600 g/ml
De 1,600 a 1,700 g/ml
De 1,700 a 1,800 g/ml
Longitud (métrico):
250 mm
270 mm
300 mm
Unit Size:
1 PCS
cada uno
This item is not returnable. View return policy
Product Code. 11788083
Brand: Alla France™ 3500MC135/20/Hqp
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
Cantidad 1Each
Tipo Hidrómetro de densidad
Exactitud ±0,001
Longitud (métrico) 270 mm
Intervalo de gravedad específica De 1,350 a 1,400 g/ml
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.