Learn More
Gilson™ Micropipetas multicanal PIPETMAN™ M
Tienda Gilson Productos
Click to view available options
Número de canales:
1
8
12
Modelo:
P10mLM
P1200M
P12X10M
P12x100M
P12x1200M
P12x200M
P12x20M
P12x300M
P300M
P5000M
P8X20M
P8x100M
P8x10M
P8x1200M
P8x200M
P8x300M
Volumen (métrico):
1 µl-10 ml
De 0,5 to 10 μl
De 1 a 20 μl
De 10 a 10 μl
De 10 a 300 μl
De 10 to 100 μl
De 100 a 1200 μl
De 20 a 300 μl
De 20 to 200 μl
De 50 to 1200 μl
De 500 to 5000 μl
Puntas compatibles:
D1000, DF1000, D1200, DF1200
D10 ml
D1200
D200
D200, DF200, D300, DF300
D5000
DF100
DF1200
Puntas de D10, DL10, DF10 o DFL10
Puntas de D10, DL10, DF10, DFL10
Puntas de D200, D300, DF100ST, DF200ST, DF300ST
Puntas de DL10, DF30ST, D200
Tamaño de la unidad:
cada uno
Descripción
- Motorized piston with adjustable piston speed that is compatible with varying solution densities
- Lockable volume to prevent inaccuracies
- Color-coded push-button corresponds to matching tip packs
- Service alert reminders
- Unique tip holder design allows for a secure fit of most brands of tips
- Features four essential pipetting modes dash (en) reverse, mix, repetitive and pipetting to best fit your application
- Pipette can be used while charging eliminating bottlenecks and downtime
- Designed to exert equal pressure on each piston to ensure consistent volume levels across all channels
- Power carrousel conveniently holds and charges up to 5 PIPETMAN M pipettes at once, charging 80% of its full capacity in less than one hour
- Gilson Trident and unique spacer distribute ejection forces evenly over each tip, reducing the force required to eject tips
Especificaciones
Especificaciones
| Puntas compatibles | Puntas de D10, DL10, DF10 o DFL10 |
| Color | Verde |
| Modelo | P12x300M |
| Número de canales | 12 |
| Línea de productos | Pipetman |
| Tipo | Pipeta |
| Volumen (métrico) | De 10 a 300 μl |
| Estilo de dedo | Ergonómico |
| Garantía | 1 año |