Learn More
Resch Glas™ Vial con borde enrollado de vidrio
Glass Rolled Rim Vials are ideal for a variety of laboratory applications. The vials are made of soda-lime glass with a tight closing polyethylene snap-on lid, non-sterile.
Marca: Resch Glas™ 9400215
Detalles adicionales : Peso : 1.36800kg
Descripción
- Soda-lime glass
- PE snap-on lid
- Non-sterile
Especificaciones
Natural | |
19 mm | |
Vidrio | |
5 ml | |
1.6 in. | |
Rolled Rim | |
Polietileno | |
40 mm | |
Rolled Rim Vial |
Rolled rim glass vials with snap-on lid | |
Transparente | |
Non-sterile | |
A variety of laboratory applications | |
Polyethylene | |
0.2 oz. | |
Tapa a presión | |
200/Paq. |