Learn More
Rogo Sampaic™ Pipetas volumétricas de vidrio de clase B.
Código de producto. 11702684
Click to view available options
Volumen (métrico):
2 ml
10 ml
20 ml
25 ml
50 ml
Color:
Amarillo
Naranja
Negro
Rojo
Unit Size:
cada uno
paquete de 2
This item is not returnable. View return policy
Product Code. 11702684
Brand: Rogo Sampaic™ DGG005
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
Provides an extremely high degree of volumetric accuracy for your pipetting needs. Rogo Sampaic™ Class B Glass Volumetric Pipettes are manufactured from clear soda lime glass.
  • Con graduaciones en ámbar vitrificado
  • Con perilla de seguridad
Specifications
Calibrado
Calibración 0,1 ml
Clase B
Graduado Sí
Color de graduación Marrón
Graduaciones 0,1 ml
Color Amarillo
Volumen (métrico) 20 ml
Longitud (métrico) 455 mm
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.