missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Human IgA Secondary Antibody, FITC, Invitrogen™
Goat Polyclonal Secondary Antibody
Marca: Invitrogen A18782
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The sensitivity of each lot of antibody is confirmed using ELISA. The specificity of each lot of antibody is confirmed by immunoelectrophoresis (IEP).
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Especificaciones
Human IgA | |
Polyclonal | |
FITC | |
Goat | |
Affinity chromatography | |
RUO | |
Human | |
Liquid |
Flow Cytometry, Immunocytochemistry | |
1.5 mg/mL | |
PBS with 1% BSA and 0.05% sodium azide; pH 7.8 | |
Human IgA | |
1 mg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido