missing translation for 'onlineSavingsMsg'
Learn More

Goat anti-Human IgG Fab, Superclonal™ Recombinant Secondary Antibody, Invitrogen™

Goat Recombinant Polyclonal Secondary Antibody

104.00€ - 254.00€

Especificaciones

Antígeno Human IgG Fab
Concentración 1.0 mg/mL
Aplicaciones ELISA, Immunocytochemistry, Western Blot
Clasificación Recombinant Polyclonal
Conjugado Unconjugated
Ver más especificaciones

Productos 2
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
17846592
Ver documentos
Invitrogen
A5685650UL
50 μL
104.00€
50 microlitros
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
17886092
Ver documentos
Invitrogen
A56856
250 μL
254.00€
250 microlitros
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
Descripción

Descripción

This Goat-anti-Human IgG Fab, Superclonal™ Recombinant Secondary Antibody detects human IgG Fab with specificity to the light chains kappa and lambda as well as the heavy chain CH1. This antibody does not detect the Fc and does not cross-react with immunoglobulins from other species. Recombinant antibodies are produced using specific genes that code for the desired antibodies. These genes are cloned into an expression vector and expressed in vitro. The advantages of recombinant antibodies include better specificity, animal origin-free formulation, and lot-to-lot consistency. Superclonal™ Recombinant antibodies are a defined pool of recombinant monoclonal antibodies, manufactured by selecting and pooling individual clones to provide complementary, specific, and optimal epitope coverage to achieve enhanced performance. The sensitivity and specificity of each lot is confirmed using ELISA. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Especificaciones

Especificaciones

Human IgG Fab
ELISA, Immunocytochemistry, Western Blot
Unconjugated
Goat
Human
Purified Human IgG
Secondary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
1.0 mg/mL
Recombinant Polyclonal
Liquid
RUO
PBS with 0.09% sodium azide; pH 7.4
IgG
Protein G
Videos
SDS
Documentos

Documentos

Product Certifications