Learn More
Invitrogen™ Goat anti-Rabbit IgG (Heavy chain), Superclonal™ Recombinant Secondary Antibody, Alexa Fluor™ Plus 647, Invitrogen™
Goat Recombinant Polyclonal Secondary Antibody
117.00€ - 351.00€
Especificaciones
Antígeno | Rabbit IgG (Heavy chain) |
---|---|
Concentración | 1 mg/mL |
Aplicaciones | Flow Cytometry, Immunocytochemistry, Western Blot |
Clasificación | Recombinant Superclonal |
Conjugado | Alexa Fluor Plus 647 |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
17699296
|
Invitrogen™
A5505550UL |
50 μL |
117.00€
50 microlitros |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
17689296
|
Invitrogen™
A55055 |
250 μL |
351.00€
250 microlitros |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
Descripción
Recombinant antibodies are produced using specific genes that code for the desired antibodies. These genes are cloned into an expression vector and expressed in vitro. The advantages of recombinant antibodies include: better specificity and superior lot-to-lot consistency. Minimal cross-reactivity with mouse, rat, human, bovine, guinea pig and donkey IgG is observed. Obtain superior images with the new Invitrogen Alexa Fluor Plus secondary antibodies. Make your low abundant targets visible, minimize time spent optimizing, and make your precious samples count. Alexa Fluor Plus secondary antibodies provide brighter signal compared to leading Alexa Fluor secondary antibodies, providing you with better signal-to-noise for your critical experiments. Alexa Fluor Plus secondary antibodies represent an advancement in fluorescent conjugate technology. Alexa Fluor Plus secondary antibodies are conjugated using new, proprietary dye chemistry so you can generate stunning images. Using conjugate solutions: Centrifuge the protein conjugate solution briefly in a microcentrifuge before use; add only the supernatant to the experiment. This step will help eliminate any protein aggregates that may have formed during storage, thereby reducing nonspecific background staining. Because staining protocols vary with application, the appropriate dilution of antibody should be determined empirically.
Anti-Rabbit secondary antibodies are affinity-purified antibodies with well-characterized specificity for rabbit immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Especificaciones
Rabbit IgG (Heavy chain) | |
Flow Cytometry, Immunocytochemistry, Western Blot | |
Alexa Fluor Plus 647 | |
Goat | |
Rabbit | |
Purified Rabbit Immunoglobulin. | |
Secondary | |
4°C, store in dark, DO NOT FREEZE! |
1 mg/mL | |
Recombinant Superclonal | |
Liquid | |
RUO | |
proprietary buffer with 0.008% Bromonitrodioxane, 0.008% Methylisothiazolone; pH 6.5 | |
IgG | |
Affinity chromatography | |
Secondary Antibody |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.