Learn More
GPR39 Antibody (189702) [Janelia Fluor™ 646], Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals FAB3348J
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
GPR39 Monoclonal antibody specifically detects GPR39 in Human samples. It is validated for Flow Cytometry, CyTOF
Especificaciones
GPR39 | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
189702 | |
Flow Cytometry, CyTOF-ready | |
G protein-coupled receptor 39, G-protein coupled receptor 39, MGC149541 | |
NS0 mouse myeloma cell line transfected with human GPR39, Met1-Val453, Accession # BAG37409 | |
0.1 mL | |
GPCR, Lipid and Metabolism | |
2863 | |
Store at 4°C in the dark. | |
IgG2a |
Título del producto
GPR39 Antibody (189702) [Janelia Fluor™ 646], Novus Biologicals™ > 0.1 mL; Janelia Fluor 646
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido