missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR50 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA533731
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey (100%) Marmoset, Panda, Dog (89%).
Melatonin-related receptor expression has been documented from human CNS tissues, specifically in hypothalamus and pituitary, but not in human peripheral tissues. In animals, expression has also been seen in the CNS, and peripheral tissues.Especificaciones
GPR50 | |
Polyclonal | |
Unconjugated | |
GPR50 | |
G protein-coupled receptor 50; Gpr50; G-protein-coupled receptor 50; H9; Mel1c; melatonin-related receptor; MTNRL | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9248 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q13585 | |
GPR50 | |
Synthetic 18 amino acid peptide from C-terminus of human GPR50. | |
50 μg | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG |