Learn More
KARTELL Graduated Bottle
The Graduated Bottle has a specific design with a rectangular base for easy storage.
Marca: KARTELL 0047900
Detalles adicionales : Peso : 0.10000kg
Descripción
- Specific design with rectangular base thought for easy storage of the bottles on the shelves and for enhanced stability when in use; the ergonomic design of the handgrip allows easy pouring.
- Screw capped and graduated up to 2000 mL.
- CE IVDR product
Especificaciones
Urine Bottle | |
Yes | |
Non-sterile | |
2 L |
Reusable | |
HDPE | |
0.53 gal. |