Learn More
GSTP1 Monoclonal Antibody (3F2C2), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA515309
328.68 EUR hasta el 2025-03-29
Use el código promocional "24111" para aplicar la oferta.
Alertas:
Para beneficiarse del descuento, los clientes deben comprar tres unidades del mismo producto al precio de catálogo en un solo pedido para recibir el 33.33% de descuento No hay límite para los múltiplos de 3 que los clientes pueden comprar PCODE:24111
Descripción
MA5-15309 targets GSTP1 in indirect ELISA, FACS, IF, IHC, and WB applications and shows reactivity with Human samples. The MA5-15309 immunogen is purified recombinant fragment of human GSTP1 expressed in E. Coli. MA5-15309 detects GSTP1 which has a predicted molecular weight of approximately 23kDa.
Glutathione S-transferases (GSTs) are a family of enzymes that play an important role in detoxification by catalyzing the conjugation of many hydrophobic and electrophilic compounds with reduced glutathione. Based on their biochemical, immunologic, and structural properties, the soluble GSTs are categorized into 4 main classes: alpha, mu, pi, and theta. This GST family member is a polymorphic gene encoding active, functionally different GSTP1 variant proteins that are thought to function in xenobiotic metabolism and play a role in susceptibility to cancer, and other diseases.
Especificaciones
GSTP1 | |
Monoclonal | |
Conc. not determined | |
ascites with 0.03% sodium azide | |
P09211 | |
Gstp1 | |
Purified recombinant fragment of human GSTP1 expressed in E. Coli. | |
RUO | |
2950 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot | |
3F2C2 | |
Unconjugated | |
Gstp1 | |
Chain 7; deafness, X-linked 7; DFN7; epididymis secretory protein Li 22; FAEES3; fatty acid ethyl ester synthase III; glutathione S-transferase P; Glutathione S-transferase P 1; glutathione S-transferase pi 1; glutathione S-transferase pi 2; glutathione S-transferase, pi 1; glutathione S-transferase, pi 2; Glutathione-S-transferase placental enzyme pi type; glutathione-S-transferase, pi 1; Glutathione-S-transferase, placental enzyme pi type; GST 7-7; GST class-pi; gst P1; Gst p-1; GST YF-YF; Gst3; Gstp; GST-P; Gstp1; GSTP1-1; Gstp2; GstpiB; GST-piB; HEL-S-22; HGNC:4638; PI; Preadipocyte growth factor | |
Mouse | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.