missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ GTF2B (Human) Recombinant Protein
Human GTF2B full-length ORF ( AAH20597, 1 a.a. - 316 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00002959-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
- Sequence: MASTSRLDALPRVTCPNHPDAILVEDYRAGDMICPECGLVVGDRVIDVGSEWRTFSNDKATKDPSRVGDSQNPLLSDGDLSTMIGKGTGAASFDEFGNSKYQNRRTMSSSDRAMMNAFKEITTMADRVNLPRNIVDRTNNLFKQVYEQKSLKGRANDAIASACLYIACRQEGVPRTFKEICAVSRISKKEIGRCFKLILKALETSVDLITTGDFMSRFCSNLCLPKQVQMAATHIARKAVELDLVPGRSPISVAAAAIYMASQASAEKRTQKEIGDIAGVADVTIRQSYRLIYPRAPDLFPTDFKFDTPVDKLPQL
Especificaciones
AAH20597 | |
general transcription factor IIB | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GTF2B | |
GST |
2959 | |
Wheat germ expression system | |
10 μg | |
TF2B, TFIIB | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |