missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ GTF2E2 (Human) Recombinant Protein
Human GTF2E2 full-length ORF ( AAH30572, 1 a.a. - 291 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00002961-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
- Sequence: MDPSLLRERELFKKRALSTPVVEKRSASSESSSSSSKKKKTKVEHGGSSGSKQNSDHSNGSFNLKALSGSSGYKFGVLAKIVNYMKTRHQRGDTHPLTLDEILDETQHLDIGLKQKQWLMTEALVNNPKIEVIDGKYAFKPKYNVRDKKALLRLLDQHDQRGLGGILLEDIEEALPNSQKAVKALGDQILFVNRPDKKKILFFNDKSCQFSVDEEFQKLWRSVTVDSMDEEKIEEYLKRQGISSMQESGPKKVAPIQRRKKPASQKKRRFKTHNEHLAGVLKDYSDITSSK
Especificaciones
AAH30572 | |
general transcription factor IIE, polypeptide 2, beta 34kDa | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GTF2E2 | |
GST |
2961 | |
Wheat germ expression system | |
25 μg | |
FE, TF2E2, TFIIE-B | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |