missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GTF3C5 Rabbit anti-Human, Polyclonal, Abnova™
Rabbit Polyclonal Antibody
Marca: Abnova PAB30123.100uL
Detalles adicionales : Peso : 0.02000kg
Descripción
Rabbit polyclonal antibody raised against synthetic peptide of human GTF3C5.
- High specificity
- High reactivity
Especificaciones
GTF3C5 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of human GTF3C5. | |
In PBS (2% sucrose, 0.09% sodium azide). | |
FLJ20857/TFIIIC63/TFIIICepsilon/TFiiiC2-63 | |
Rabbit | |
Affinity purification | |
RUO | |
9328 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (4-8 ug/mL) Western Blot (0.2-1 ug/mL) The optimal working dilution should be determined by the end user. | |
NM_012087 | |
GTF3C5 | |
A synthetic peptide corresponding to N-terminus of human GTF3C5. | |
100 μL | |
Primary | |
Human | |
Liquid |
For Research Use Only (RUO)