missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Woodsafe Hazardous Waste Bin - Sharpsbin
A cost-effective solution for lowering the CO2 emissions. Eco-friendly sharps and hazardous waste containers, produced from renewable resources using green energy. Reducing CO2 emissions by up to 66%, documented in a full LCA. Available in sizes from 0.5 L to 50 L.
2462.30€ - 2775.76€
Especificaciones
Certificaciones/conformidad | EN ISO 23907-1:2019. UN3291, ADR Approved |
---|---|
Tipo de tapón | Temporary and permanent closure |
Color del tapón | Yellow |
Descripción | Environmental friendly Sharpsbin |
Color | Brown, Yellow |
Alertas:
Follow instructions for safe and correct use
Product Code | Brand | Capacidad (métrico) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacidad (métrico) | Price | Quantity & Availability | |||||
18752825
|
Woodsafe
1250222 |
25 L |
2462.30€
paquete de 132 |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
18712825
|
Woodsafe
1500222 |
50 L |
2775.76€
paquete de 96 |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
Description
- Woodsafe™ offer a sustainable clinical waste management solution produced with residue from the forest industry
- Lowers the carbon footprint by more than 60%, proved in full LCA
- Made from 100% Biocomposite
- The lid can be temporarily closed to conceal the contents when not in use and securely permanently closed for transportation to the final disposal
- Natural colour and smell from wood - no migration of substances - EN - tested for toys and food
- Compliant with UN3291
- Tested according to ISO 23907-1:2019
- UN ADR Approved
- Biocomposite have the same properties as fossil-based plastic and can be used exactly the same way
- NOTE: String between lids is for delivery purpose, remove/break before use!
- Stop burning plastic
Specifications
EN ISO 23907-1:2019. UN3291, ADR Approved | |
Yellow | |
Brown, Yellow | |
Sí | |
Autoclavable | |
Safe Disposal of sharps and potentially Infectious Waste or medical waste | |
Biocomposite |
Temporary and permanent closure | |
Environmental friendly Sharpsbin | |
Single-use | |
No estéril | |
Bin + Lid + Label | |
Pallet | |
Biohazard Waste Container (Benchtop) |
Lower climate impact
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title