missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ HBsAg (adr) (ACX36042, 227 amino acids) Full-length Recombinant Protein expressed in CHO cells
Used for SDS-PAGE
Marca: Abnova™ P4874.50ug
Detalles adicionales : Peso : 0.02000kg
Descripción
Sequence: MENTTSGFLGPLLVLQAGFFLLTRILTIPQSLDSWWTSLNFLGGAPTCPGQNSQSPTSNHSPTSCPPICPGYRWMCLRRFIIFLFILLLCLIFLLVLLDYQGMLPVCPLLPGTSTTSTGPCKTCTIPAQGTSMFPSCCCTKPSDGNCTCIPIPSSWAFARFLWEWASVRFSWLSLLVPFVQWFVGLSPTVWLSVIWMMWYWGPSLYNILSPFLPLLPIFFCLWVYIEspecificaciones
ACX36042 | |
Liquid | |
HBsAg (adr) Recombinant Protein | |
Proprietary chromatography | |
MENTTSGFLGPLLVLQAGFFLLTRILTIPQSLDSWWTSLNFLGGAPTCPGQNSQSPTSNHSPTSCPPICPGYRWMCLRRFIIFLFILLLCLIFLLVLLDYQGMLPVCPLLPGTSTTSTGPCKTCTIPAQGTSMFPSCCCTKPSDGNCTCIPIPSSWAFARFLWEWASVRFSWLSLLVPFVQWFVGLSPTVWLSVIWMMWYWGPSLYNILSPFLPLLPIFFCLWVYI | |
RUO | |
Hepatitis B | |
None | |
Liquid |
SDS-PAGE | |
24kDa | |
Mammalian cell (CHO) expression system | |
50 ug | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
HBSAG | |
Recombinant | |
Mammalian cell (CHO) expression system | |
>95% by SDS-PAGE |