missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ HBsAg preS2 (AAK51534, 55 amino acids) Recombinant Protein
Used for SDS-PAGE
Marca: Abnova™ P4877.50ug
Detalles adicionales : Peso : 0.02000kg
Description
Sequence: MQWNSTTFHQALLDPKVRGLYFPAGGSSSGTVNPVPTTASPISSIFSRTGDPAPNSpécification
AAK51534 | |
Liquid | |
HBsAg (preS2) Recombinant Protein | |
Proprietary chromatography | |
MQWNSTTFHQALLDPKVRGLYFPAGGSSSGTVNPVPTTASPISSIFSRTGDPAPN | |
RUO | |
E. coli | |
None | |
Liquid |
SDS-PAGE | |
5.7kDa | |
Escherichia coli expression system | |
50 ug | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
HBSAG | |
Recombinant | |
Escherichia coli expression system | |
≥95% by SDS-PAGE |