Learn More
Invitrogen™ HCN3 Monoclonal Antibody (TLL6C5)

Descripción
MA3-902 detects HCN3 from human, mouse and rat samples. MA3-902 has been successfully used in Western blotting and immunohistochemistry procedures. By Western blot MA3-902 detects a ~90 KDa band representing HCN3. The MA3-902 immunogen is a synthetic peptide corresponding to residues T(640) L L A R S A R R S A G S P A S P L V P V R A G P L L A R G P W A S T S(675) of rat HCN3. MA3-902 (clone TLL6C5) is a rat monoclonal hybridoma, produced by immunizing a rat, isolating the spleen cells, creating the hybridoma and successively injecting these cells into a mouse to produce ascites.
Especificaciones
Especificaciones
| Antígeno | HCN3 |
| Aplicaciones | Immunohistochemistry (Paraffin), Western Blot |
| Clasificación | Monoclonal |
| Clon | TLL6C5 |
| Concentración | Conc. Not Determined |
| Conjugado | Unconjugated |
| Formulación | Ascites with 0.05% sodium azide |
| génica | Hcn3 |
| N.º de referencia del gen | O88705, Q9JKA8, Q9P1Z3 |
| Alias de gen | Bcng4; BCNG-4; Hac3; HAC-3; HCN3; Hcn4; hyperpolarization activated cyclic nucleotide gated potassium channel 3; hyperpolarization activated cyclic nucleotide-gated potassium channel 3; hyperpolarization-activated cation channel 3; hyperpolarization-activated cyclic nucleotide-gated potassium channel 3; hyperpolarization-activated, cyclic nucleotide-gated K+ 3; hyperpolarization-activated, cyclic nucleotide-gated K+ 4; KIAA1535; MGC131493; Potassium/sodium hyperpolarization-activated cyclic nucleotide-gated channel 3 |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.