Learn More
HENMT1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA528543
Detalles adicionales : Peso : 0.01000kg
Descripción
Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji. Predicted reactivity: Rhesus Monkey (99%), Chimpanzee (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
HENMT1 Polyclonal antibody specifically detects HENMT1 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Especificaciones
HENMT1 | |
Polyclonal | |
Unconjugated | |
HENMT1 | |
4921515J06Rik; C1orf59; HEN1; HEN1 methyltransferase homolog 1; HEN1 methyltransferase homolog 1 (Arabidopsis); Henmt1; hua enhancer 1 homolog 1; mHEN1; RGD1306230; Small RNA 2'-O-methyltransferase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
113802 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q5T8I9 | |
HENMT1 | |
Recombinant fragment corresponding to a region within amino acids 1 and 252 of Human LOC113802 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |