missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HEY1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 19929-1-AP-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Hairy/enhancer-of-split related with YRPW motif protein 1 belongs to the HEY family and is a Notch Signaling Pathway Protein. It contains one basic helix-loop-helix (bHLH) domain and one Orange domain. It is a downstream effector of Notch signaling which may be required for cardiovascular development. It acts as transcriptional repressor which binds preferentially to the canonical E box sequence 5′-CACGTG-3′. It represses transcription by the cardiac transcriptional activators GATA4 and GATA6. Hey1 is also reported to be expressed during development of the nervous system.Especificaciones
| HEY1 | |
| Polyclonal | |
| Unconjugated | |
| HEY1 | |
| BHLHb31, CHF 2, CHF2, HERP2, HESR 1, HESR1, HEY1, hHRT1, HRT 1, HRT1, OAF1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 15213, 155437, 23462 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot | |
| 0.31 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9WV93, Q9Y5J3 | |
| HEY1 | |
| HEY1 Fusion Protein Ag13906 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido