missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSPA7 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA535359
Detalles adicionales : Peso : 0.01000kg
Descripción
HSPA7 Polyclonal antibody specifically detects HSPA7 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot.Especificaciones
HSPA7 | |
Polyclonal | |
Unconjugated | |
HSPA7 | |
Heat shock protein; heat shock protein family A (Hsp70) member 7; HSP; HSP70B; HSPA7 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
3311 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
P48741 | |
HSPA7 | |
KLH conjugated synthetic peptide between 169-196 amino acids from the central region of human HSPA7 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |