Learn More
HTR2B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen 720256
Detalles adicionales : Peso : 0.01000kg
Description
These Polyclonal antibodies are of rabbit origin developed by immunizing animals with proteins or peptides. The polyclonal antibody is purified by affinity purification from the rabbit sera generated after immunizing the rabbits with a specific type of protein or peptide. The purified antibody is tested for its functionality in various relevant research applications. The antibody is developed for Research Use Only and is non-hazardous or non-infectious in nature. This antibody is predicted to react with Horse, Pig and Cat.
HTR2B is a g-protein coupled receptor for 5-hydroxytryptamine (serotonin). It functions as a receptor for various ergot alkaloid derivatives and psychoactive substances. Ligand binding causes a conformation change that triggers signaling via guanine nucleotide-binding proteins (G proteins) and modulates the activity of down-stream effectors. HTR2B also plays a role in perception of pain, regulation of behavior. It is required to normal proliferation of embryonic cardiac myocytes and normal heart development.Specifications
HTR2B | |
Polyclonal | |
Unconjugated | |
HTR2B | |
5-HT 2B receptor; 5-HT(2B); 5-HT2B; 5-HT-2B; 5HT2B Receptor; 5-HT2b receptor; 5-HT-2F; 5-hydroxytryptamine (serotonin) receptor 2B; 5-hydroxytryptamine (serotonin) receptor 2B, G protein-coupled; 5-hydroxytryptamine 2B receptor; 5-hydroxytryptamine receptor 2B; 5-hydroxytryptamine receptor 2B variant b; AJ012488; AV377389; HTR2B; NP75 protein; serotonin receptor 2B; Srl; Stomach fundus serotonin receptor | |
Rabbit | |
Affinity chromatography | |
RUO | |
3357 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 0.1% BSA, 30% glycerol and 0.09% sodium azide; pH 7.2 | |
P41595 | |
HTR2B | |
Peptides corresponding to Human HTR2B (aa 285-304, 468-481) | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |