missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HTR3A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA130207
Detalles adicionales : Peso : 0.01000kg
Descripción
A suggested positive control for this product is human brain tissue.
The product of this gene belongs to the ligand-gated ion channel receptor superfamily. This gene encodes subunit A of the type 3 receptor for 5-hydroxytryptamine, a biogenic hormone that functions as a neurotransmitter, a hormone, and a mitogen. This receptor causes fast, depolarizing responses in neurons after activation. It appears that the heteromeric combination of A and B subunits is necessary to provide the full functional features of this receptor, since either subunit alone results in receptors with very low conductance and response amplitude. Alternatively spliced transcript variants encoding different isoforms have been identified.Especificaciones
HTR3A | |
Polyclonal | |
Unconjugated | |
Htr3a | |
5HT3; 5-HT3; 5-HT-3; 5-HT3 receptor; 5HT3 serotonin receptor; 5-HT3A; 5-HT3-A; 5HT3R; 5-HT3R; 5-hydroxytryptamine (serotonin) receptor 3A; 5-hydroxytryptamine (serotonin) receptor 3A, ionotropic; 5-hydroxytryptamine receptor 3; 5-hydroxytryptamine receptor 3A; Htr3; Htr3a; serotonin receptor; Serotonin receptor 3A; serotonin-gated ion channel receptor; truncated receptor | |
Rabbit | |
Protein G | |
RUO | |
15561, 3359, 79246 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.05% sodium azide | |
P23979, P35563, P46098 | |
Htr3a | |
This antibody was developed by immunizing rabbits with a mixture of synthetic peptides corresponding to amino acids 27-42 (RATQAHSTTQPALLRL) and 427-442 (LSSIRHSLEKRDEMRE) of rat 5-HT3 Receptor, accession number P35563 | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |