Learn More
Abnova™ Human ADAMTS5 amino acids 625-930 deleted Recombinant Protein with His tag expressed in insect cells
Used for Func, SDS-PAGE
Marca: Abnova™ P4931.5ug
Detalles adicionales : Peso : 0.02000kg
Descripción
This gene encodes a member of the ADAMTS (a disintegrin and metalloproteinase with thrombospondin motifs) protein family. Members of the family share several distinct protein modules, including a propeptide region, a metalloproteinase domain, a disintegrin-like domain, and a thrombospondin type 1 (TS) motif. Individual members of this family differ in the number of C-terminal TS motifs, and some have unique C-terminal domains. The enzyme encoded by this gene contains two C-terminal TS motifs and functions as aggrecanase to cleave aggrecan, a major proteoglycan of cartilage. [provided by RefSeq]
Especificaciones
Q9UNA0 | |
Functional Study, SDS-PAGE | |
11096 | |
ADAMTS5 (d625-930) (Human) Recombinant Protein | |
15% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ADAMTS11/ADMP-2/FLJ36738 | |
ADAMTS5 | |
Baculovirus | |
His | |
Liquid |
0.1 mg/mL | |
Liquid | |
40.8kDa | |
Insect cell expression system | |
5 ug | |
RUO | |
ADAMTS5 | |
≥ 5mU/mg | |
Recombinant | |
Insect cell expression system | |
≥50% by SDS-PAGE |